Order Entry
Germany
ContactUsLinkComponent
4-Iodo-2-nitroanisole
4-Iodo-2-nitroanisole
Catalog # APOSOR400184-5G
Supplier:  Apollo Scientific
CAS Number:  
4-Iodo-2-nitroanisole
Catalog # APOSOR400184-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :279.03 g/mol
Storage Temperature:Ambient
CAS Number:52692-09-8
Formula:C₇H₆INO₃
MDL Number:MFCD08060933