Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-3-methoxybenzaldehyde
2-Bromo-3-methoxybenzaldehyde 95%
2-Bromo-3-methoxybenzaldehyde 95%
Irritant
2-Bromo-3-methoxybenzaldehyde 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Bromo-3-methoxybenzaldehyde 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
10401-18-0
:
Formula:
C₈H₇BrO₂
MDL Number:
MFCD09834663
:
215.04 g/mol
Melting Pt:
67…69 °C
:
GHS07
Storage Temperature:
Ambient