Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Isoquinoline-5-amine
Isoquinoline-5-amine
Isoquinoline-5-amine
Irritant
Isoquinoline-5-amine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Isoquinoline-5-amine
Supplier:
Apollo Scientific
About this item
CAS Number:
1125-60-6
:
Formula:
C₉H₈N₂
MDL Number:
MFCD00006907
Melting Pt:
125…131 °C
:
144.18 g/mol
EINECS:
214-408-3
:
GHS07
Storage Temperature:
Refrigerator