Order Entry
Germany
ContactUsLinkComponent
1-Boc-6-chloroindole-3-boronic acid pinacol ester 98%
1-Boc-6-chloroindole-3-boronic acid pinacol ester 98%
Catalog # APOSOR360720-250MG
Supplier:  Apollo Scientific
CAS Number:  
1-Boc-6-chloroindole-3-boronic acid pinacol ester 98%
Catalog # APOSOR360720-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :377.6701 g/mol
Storage Temperature:Ambient
CAS Number:1218790-24-9
Formula:C₁₉H₂₅BClNO₄
MDL Number:MFCD15143606