1-Boc-5-chloroindole-3-boronic acid pinacol ester 95+%
Catalog # APOSOR360719-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:377.6701 g/mol
Storage Temperature:Ambient
CAS Number:1218790-30-7
Formula:C₁₉H₂₅BClNO₄
MDL Number:MFCD15143612