Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dibenzothiophene-2-boronic acid
Dibenzothiophene-2-boronic acid 99%
Dibenzothiophene-2-boronic acid 99%
Irritant
Dibenzothiophene-2-boronic acid 99%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Dibenzothiophene-2-boronic acid 99%
Supplier:
Apollo Scientific
About this item
CAS Number:
668983-97-9
:
Formula:
C₁₂H₉BO₂S
MDL Number:
MFCD01318982
:
228.07 g/mol
:
GHS07
Storage Temperature:
Freezer