Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-5-(methylthio)pyridine
3-Bromo-5-(methylthio)pyridine 95%
3-Bromo-5-(methylthio)pyridine 95%
Irritant
3-Bromo-5-(methylthio)pyridine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Bromo-5-(methylthio)pyridine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
142137-18-6
:
Formula:
C₆H₆BrNS
MDL Number:
MFCD08689537
:
GHS07
Storage Temperature:
Ambient