2-sec-Butyl-3-methoxypyrazine
Catalog # APOSOR350347-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:166.2203 g/mol
Storage Temperature:Ambient
CAS Number:24168-70-5
Formula:C₉H₁₄N₂O
MDL Number:MFCD00006136