Order Entry
Germany
ContactUsLinkComponent
(R)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane
(R)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane
Catalog # APOSOR350338-25G
Supplier:  Apollo Scientific
CAS Number:  
(R)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane
Catalog # APOSOR350338-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :150.60 g/mol
Storage Temperature:Ambient
CAS Number:57044-24-3
Formula:C₆H₁₁ClO₂
MDL Number:MFCD00273371