3,5-Dihydroxytoluene monohydrate
Catalog # APOSOR350224-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:142.15 g/mol
Storage Temperature:Ambient
CAS Number:6153-39-5
Formula:C₇H₈O₂·1H₂O
MDL Number:MFCD00149092