

Methyl-4-iodobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:619-44-3
:
Formula:C₈H₇IO₂
MDL Number:MFCD00016353
Melting Pt:112…116 °C
Storage Temperature:Ambient
EINECS:210-597-1
UN:3077
ADR:9+EHS,III
:262.05 g/mol
:GHS07
:GHS09
Frequently Bought Together