Order Entry
Germany
ContactUsLinkComponent
2,4-Di-tert-butyl-5-nitrophenol 98%
2,4-Di-tert-butyl-5-nitrophenol 98%
Catalog # APOSOR350138-1G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Di-tert-butyl-5-nitrophenol 98%
Catalog # APOSOR350138-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :251.33 g/mol
Storage Temperature:Ambient
CAS Number:873055-57-3
Formula:C₁₄H₂₁NO₃
MDL Number:MFCD11052606