1-Boc-5,6-dihydro-2H-pyridine-3-boronic acid pinacol ester
Catalog # APOSOR350037-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:309.22 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:885693-20-9
Formula:C₁₆H₂₈BNO₄
MDL Number:MFCD10697911