Boc-L-tyrosine methyl ester
Catalog # APOSOR350024-500G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:295.3 g/mol
Melting Pt:100…104 °C
:GHS07
Storage Temperature:Ambient
CAS Number:4326-36-7
Formula:C₁₅H₂₁NO₅
MDL Number:MFCD00191181