Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Phenyl-2-thiouracil
6-Phenyl-2-thiouracil
6-Phenyl-2-thiouracil
6-Phenyl-2-thiouracil
Supplier:
Apollo Scientific
Certificates
Expand image
6-Phenyl-2-thiouracil
Supplier:
Apollo Scientific
About this item
CAS Number:
36822-11-4
:
Formula:
C₁₀H₈N₂OS
MDL Number:
MFCD00059166
:
204.25 g/mol
Storage Temperature:
Ambient