4-Acetylbenzoic acid 96%
Catalog # APOSOR345603-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:164.16 g/mol
Melting Pt:206…210 °C
EINECS:209-558-5
:GHS07
Storage Temperature:Ambient
CAS Number:586-89-0
Formula:C₉H₈O₃
MDL Number:MFCD00002561