Sodium bis(trimethylsilyl)amide 2 M in THF
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1070-89-9
:
Formula:C₆H₁₈NNaSi₂
MDL Number:MFCD00009835
:183.38 g/mol
Density:0.92 g/cm³ (25 °C)
Flash Pt:–17 °C
UN:2924
ADR:3+8,II
:GHS02
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient