Order Entry
Germany
ContactUsLinkComponent
1-(2-Phenylpyrazolo[1,5-a]pyridin-3-yl)-1-ethanone 95%
1-(2-Phenylpyrazolo[1,5-a]pyridin-3-yl)-1-ethanone 95%
Catalog # APOSOR33583-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Phenylpyrazolo[1,5-a]pyridin-3-yl)-1-ethanone 95%
Catalog # APOSOR33583-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :236.28 g/mol
Storage Temperature:Ambient
CAS Number:122643-81-6
Formula:C₁₅H₁₂N₂O
MDL Number:MFCD09759184