Order Entry
Germany
ContactUsLinkComponent
tert-Butyl-2-(2-chloroacetyl)-1,2-diazepane-1-carboxylate 95%
tert-Butyl-2-(2-chloroacetyl)-1,2-diazepane-1-carboxylate 95%
Catalog # APOSOR33490-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-2-(2-chloroacetyl)-1,2-diazepane-1-carboxylate 95%
Catalog # APOSOR33490-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :276.77 g/mol
Storage Temperature:Ambient
CAS Number:1135283-02-1
Formula:C₁₂H₂₁ClN₂O₃
MDL Number:MFCD11840976