Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-4,5-dibromo-2-thiophenecarboxylate
Methyl-4,5-dibromo-2-thiophenecarboxylate 95%
Methyl-4,5-dibromo-2-thiophenecarboxylate 95%
Irritant
Methyl-4,5-dibromo-2-thiophenecarboxylate 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl-4,5-dibromo-2-thiophenecarboxylate 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
62224-24-2
:
Formula:
C₆H₄Br₂O₂S
MDL Number:
MFCD00831688
:
299.98 g/mol
Melting Pt:
77…81 °C
:
GHS07
Storage Temperature:
Ambient