Methyl-1-(4-chlorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:338748-81-5
:
Formula:C₁₅H₁₆ClNO₃
MDL Number:MFCD00975047
:293.75 g/mol
Storage Temperature:Ambient