Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Boc-glycine (Boc-Gly-OH)
N-Boc-glycine (boc-gly-OH) 97%
N-Boc-glycine (boc-gly-OH) 97%
N-Boc-glycine (boc-gly-OH) 97%
Supplier:
Apollo Scientific
Certificates
Expand image
N-Boc-glycine (boc-gly-OH) 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
4530-20-5
:
Formula:
C₇H₁₃NO₄
MDL Number:
MFCD00002690
:
175.2 g/mol
Melting Pt:
87…90 °C
:
GHS05
:
GHS07
Storage Temperature:
Refrigerator