Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Methylmorpholine N-oxide
N-Methylmorpholine-N-oxide
N-Methylmorpholine-N-oxide
N-Methylmorpholine-N-oxide
Supplier:
Apollo Scientific
Certificates
Expand image
N-Methylmorpholine-N-oxide
Supplier:
Apollo Scientific
About this item
CAS Number:
7529-22-8
:
Formula:
C₅H₁₁NO₂
MDL Number:
MFCD00005947
:
117.15 g/mol
Melting Pt:
180…184 °C
EINECS:
231-391-8
:
GHS07
Storage Temperature:
Refrigerator
Related Information
Documentation
Certificates
Search by Lot Number
Documentation