Order Entry
Germany
ContactUsLinkComponent
N-Succinimidyl 4-Formylbenzoate
N-Succinimidyl 4-Formylbenzoate
Catalog # APOSOR323071-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Succinimidyl 4-Formylbenzoate
Catalog # APOSOR323071-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :247.21 g/mol
Storage Temperature:Ambient
CAS Number:60444-78-2
Formula:C₁₂H₉NO₅
MDL Number:MFCD00133733