5-(1-Pyrrolidinyl)-2-furaldehyde
Catalog # APOSOR322455-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:165.19 g/mol
Storage Temperature:Ambient
CAS Number:84966-28-9
Formula:C₉H₁₁NO₂
MDL Number:MFCD02629619