Order Entry
Germany
Orders LinkContactUsLinkComponent
3-Bromophenetole

3-Bromophenetole

New Product
Supplier:   Apollo Scientific
3-Bromophenetole
Supplier:  Apollo Scientific
New Product
Certificates
3-Bromophenetole

5 Options Available Below

Product Details & Documents
Synonyms:  3-Bromophenyl ethyl ether, 1-Bromo-3-ethoxybenzene
  • CAS Number:2655-84-7
  • Formula:C₈H₉BrO
  • MDL Number:MFCD00274287
  • Molecular Weight:201.06 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 33,30 €
    33,30 €
    1 * 5 g
       33,30 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 106,00 €
    106,00 €
    1 * 25 g
       106,00 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 238,00 €
    238,00 €
    1 * 100 g
       238,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 140,00 €
    1 140,00 €
    1 * 500 g
       1 140,00 €

    Specifications:
    New Product
    New Product