Order Entry
Germany
ContactUsLinkComponent
1,6-Hexanediol 97%
1,6-Hexanediol 97%
Catalog # APOSOR322268-250G
Supplier:  Apollo Scientific
CAS Number:  
1,6-Hexanediol 97%
Catalog # APOSOR322268-250G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :118.18 g/mol
Boiling Pt:248…250 °C
Melting Pt:40…43 °C
Density:1.12 g/cm³ (25 °C)
 :GHS07
Storage Temperature:Ambient
CAS Number:629-11-8
Formula:C₆H₁₄O₂
MDL Number:MFCD00002985