
1-Bromo-4-methylcyclohexane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6294-40-2
:
Formula:C₇H₁₃Br
MDL Number:MFCD00014282
:177.08 g/mol
Boiling Pt:94…96 °C/50mmHg
Density:1.262 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient