Triethylammonium chloride
Catalog # APOSOR322028-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:137.65 g/mol
Melting Pt:254…261 °C (dec.)
Storage Temperature:Ambient
CAS Number:554-68-7
Formula:C₆H₁₆ClN
MDL Number:MFCD00012500