(2R,4S)-N-Boc-4-hydroxypiperidine-2-carboxylic acidmethyl ester
Catalog # APOSOR321526-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:259.302 g/mol
Storage Temperature:Refrigerator
CAS Number:321744-26-7
Formula:C₁₂H₂₁NO₅
MDL Number:MFCD02259744