Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl-4-acetylpiperidine-1-carboxylate
tert-Butyl-4-acetylpiperidine-1-carboxylate 98%
tert-Butyl-4-acetylpiperidine-1-carboxylate 98%
tert-Butyl-4-acetylpiperidine-1-carboxylate 98%
Supplier:
Apollo Scientific
Certificates
Expand image
tert-Butyl-4-acetylpiperidine-1-carboxylate 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
206989-61-9
:
Formula:
C₁₂H₂₁NO₃
MDL Number:
MFCD08437655
:
227.3000 g/mol
:
GHS07
Storage Temperature:
Ambient