Order Entry
Germany
ContactUsLinkComponent
tert-Butyl-4-((methylamino)methyl)piperidine-1-carboxylate 97%
tert-Butyl-4-((methylamino)methyl)piperidine-1-carboxylate 97%
Catalog # APOSOR321451-5G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-4-((methylamino)methyl)piperidine-1-carboxylate 97%
Catalog # APOSOR321451-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :228.3300 g/mol
Storage Temperature:Ambient
CAS Number:138022-02-3
Formula:C₁₂H₂₄N₂O₂
MDL Number:MFCD06656629