N,N,3-Trimethylazetidin-3-amine hydrochloride
Catalog # APOSOR317229-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:150.65 g/mol
Storage Temperature:Ambient
CAS Number:132771-10-9
Formula:C₆H₁₅ClN₂
MDL Number:MFCD22123304