Order Entry
Germany
ContactUsLinkComponent
tert-Butyl-7-azaspiro[3.5]nonan-2-ylcarbamate 97%
tert-Butyl-7-azaspiro[3.5]nonan-2-ylcarbamate 97%
Catalog # APOSOR317096-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-7-azaspiro[3.5]nonan-2-ylcarbamate 97%
Catalog # APOSOR317096-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :240.3400 g/mol
Storage Temperature:Ambient
CAS Number:147611-03-8
Formula:C₁₃H₂₄N₂O₂
MDL Number:MFCD12198553