Order Entry
Germany
ContactUsLinkComponent
N-Boc-L-phenylalaninol
N-Boc-L-phenylalaninol
Catalog # APOSOR316043-100G
Supplier:  Apollo Scientific
CAS Number:  
N-Boc-L-phenylalaninol
Catalog # APOSOR316043-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :251.3 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:66605-57-0
Formula:C₁₄H₂₁NO₃
MDL Number:MFCD00076976