Order Entry
Germany
ContactUsLinkComponent
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl-p-toluenesulphonate
Irritant
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl-p-toluenesulphonate
Catalog # APOSOR315279-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl-p-toluenesulphonate
Catalog # APOSOR315279-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :GHS07
Boiling Pt:125 °C/ 0.5mmHg
Melting Pt:29 °C
Density:1.208 g/cm³ (25 °C)
Flash Pt:113 °C
Storage Temperature:Refrigerator
 :286.34 g/mol
CAS Number:23735-43-5
Formula:C₁₃H₁₈O₅S
MDL Number:MFCD00063234