Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Di-tert-butylcarbazole
3,6-Di-tert-butylcarbazole 98%
3,6-Di-tert-butylcarbazole 98%
Irritant
3,6-Di-tert-butylcarbazole 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,6-Di-tert-butylcarbazole 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
37500-95-1
:
Formula:
C₂₀H₂₅N
MDL Number:
MFCD03425849
:
279.43 g/mol
Melting Pt:
232 °C
:
GHS07
Storage Temperature:
Ambient