Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Diamino-6-methoxypyridine dihydrochloride
2,3-Diamino-6-methoxypyridine dihydrochloride 98%
2,3-Diamino-6-methoxypyridine dihydrochloride 98%
Irritant
2,3-Diamino-6-methoxypyridine dihydrochloride 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,3-Diamino-6-methoxypyridine dihydrochloride 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
94166-62-8
:
MFCD00137799
Formula:
C₆H₁₁Cl₂N₃O
:
212.074 g/mol
:
GHS07
Storage Temperature:
Ambient