2-Formyl-5-methoxypyridine
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 5-Methoxypyridine-2-carbaldehyde, 5-Methoxypicolinaldehyde
- CAS Number:22187-96-8
- Formula:C₇H₇NO₂
- MDL Number:MFCD08234867
- Molecular Weight:137.1360 g/mol
- Storage Temperature:Ambient
5 Options Available Below
Product Details & Documents
Synonyms: 5-Methoxypyridine-2-carbaldehyde, 5-Methoxypicolinaldehyde
- CAS Number:22187-96-8
- Formula:C₇H₇NO₂
- MDL Number:MFCD08234867
- Molecular Weight:137.1360 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



