2,5-Diazaspiro[3.4]octane-5-carboxylic acid tert-butyl ester
Catalog # APOSOR312236-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:212.29 g/mol
Storage Temperature:Ambient
CAS Number:1086398-04-0
Formula:C₁₁H₂₀N₂O₂
MDL Number:MFCD11501186