Order Entry
Germany
ContactUsLinkComponent
tert-Butyl-3-(benzylamino)-3-(2-ethoxy-2-oxoethyl)azetidine-1-carboxylate
tert-Butyl-3-(benzylamino)-3-(2-ethoxy-2-oxoethyl)azetidine-1-carboxylate
Catalog # APOSOR312050-250MG
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-3-(benzylamino)-3-(2-ethoxy-2-oxoethyl)azetidine-1-carboxylate
Catalog # APOSOR312050-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :348.45 g/mol
Storage Temperature:Ambient
CAS Number:1373923-04-6
Formula:C₁₉H₂₈N₂O₄
MDL Number:MFCD22199157