Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-4-mercaptobenzoate
Methyl-4-mercaptobenzoate
Methyl-4-mercaptobenzoate
Methyl-4-mercaptobenzoate
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl-4-mercaptobenzoate
Supplier:
Apollo Scientific
About this item
CAS Number:
6302-65-4
:
Formula:
C₈H₈O₂S
MDL Number:
MFCD01863423
:
168.22 g/mol
Melting Pt:
33…36 °C
Storage Temperature:
Refrigerator