
tert-Butyl-2-chloro-7,8-dihydro-1,6-naphthyridine-6(5H)-carboxylate 95%
Catalog # APOSOR310926-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:268.75 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1151665-15-4
Formula:C₁₃H₁₇ClN₂O₂
MDL Number:MFCD10565958