Order Entry
Germany
ContactUsLinkComponent
{2-[3-(4-Bromophenyl)-1,2,4-oxadiazol-5-yl]ethyl}amine hydrochloride
{2-[3-(4-Bromophenyl)-1,2,4-oxadiazol-5-yl]ethyl}amine hydrochloride
Catalog # APOSOR310744-1G
Supplier:  Apollo Scientific
CAS Number:  
{2-[3-(4-Bromophenyl)-1,2,4-oxadiazol-5-yl]ethyl}amine hydrochloride
Catalog # APOSOR310744-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :304.58 g/mol
Storage Temperature:Ambient
CAS Number:937666-44-9
Formula:C₁₀H₁₁BrClN₃O
MDL Number:MFCD28023529