tert-Butyl-5-bromoindoline-1-carboxylate
Catalog # APOSOR310451-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Melting Pt:125…127 °C
:298.18 g/mol
Storage Temperature:Ambient
CAS Number:261732-38-1
Formula:C₁₃H₁₆BrNO₂
MDL Number:MFCD08059280