Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Boc-indoline-5-boronic acid pinacol ester
1-Boc-indoline-5-boronic acid pinacol ester
1-Boc-indoline-5-boronic acid pinacol ester
1-Boc-indoline-5-boronic acid pinacol ester
APOSOR310450-1G
:
Apollo Scientific
:
1-Boc-indoline-5-boronic acid pinacol ester
APOSOR310450-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
120…121 °C
:
345.246 g/mol
:
Ambient
:
837392-67-3
:
C₁₉H₂₈BNO₄
:
MFCD12408237