Order Entry
Germany
Orders LinkContactUsLinkComponent
6-Bromoquinaldine

6-Bromoquinaldine

Bottle, 5 g
Catalog # APOSOR3097-5G
Corrosive
Irritant
Health
Supplier:   Apollo Scientific
6-Bromoquinaldine
Catalog # APOSOR3097-5G
Supplier:  Apollo Scientific
CAS Number:  877-42-9
6-Bromoquinaldine

Bottle, 5 g

CAS Number:  877-42-9
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:877-42-9
  • Formula:C₁₀H₈BrN
  • MDL Number:MFCD00079724
  • Molecular Weight:222.08 g/mol
  • Melting Pt:100…105 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.