Order Entry
Germany
ContactUsLinkComponent
6-Bromo-2-chloro-4-methylquinoline
6-Bromo-2-chloro-4-methylquinoline
Catalog # APOSOR309381-250MG
Supplier:  Apollo Scientific
CAS Number:  
6-Bromo-2-chloro-4-methylquinoline
Catalog # APOSOR309381-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :256.5300 g/mol
Melting Pt:138…140 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:3913-19-7
Formula:C₁₀H₇BrClN
MDL Number:MFCD06254262