Order Entry
Germany
Orders LinkContactUsLinkComponent
 

3-(2-Bromophenyl)propionic acid 98%

Irritant
New Product
Supplier:   Apollo Scientific
3-(2-Bromophenyl)propionic acid 98%
Supplier:  Apollo Scientific
New Product
Certificates
3-(2-Bromophenyl)propionic acid 98%

5 Options Available Below

Product Details & Documents
  • CAS Number:15115-58-9
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD01310791
  • Molecular Weight:229.07 g/mol
  • Melting Pt:98…102 °C
  • Storage Temperature:Ambient
  • UN:3335
  • ADR:9,III
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 79,60 €
    79,60 €
    1 * 100 g
       79,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 393,00 €
    393,00 €
    1 * 500 g
       393,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 1 960,00 €
    1 960,00 €
    1 * 2,5 kg
       1 960,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.