Order Entry
Germany
Orders LinkContactUsLinkComponent
 

cis-4-Hydroxy-L-(-)-proline 97%

Irritant
Supplier:   Apollo Scientific
cis-4-Hydroxy-L-(-)-proline 97%
Supplier:  Apollo Scientific
Certificates
cis-4-Hydroxy-L-(-)-proline 97%

6 Options Available Below

Product Details & Documents
Synonyms:  L(-)-cis-4-Hydroxyproline, H-cis-Hyp-OH, (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, (S)-(-)-cis-4-Hydroxyproline
  • CAS Number:618-27-9
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00064319
  • Molecular Weight:131.13 g/mol
  • Melting Pt:255…257 °C
  • Storage Temperature:Ambient
  • EINECS:210-542-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 21,80 €
    21,80 €
    1 * 250 mg
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 24,60 €
    24,60 €
    1 * 1 g
       24,60 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 30,40 €
    30,40 €
    1 * 5 g
       30,40 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 92,70 €
    92,70 €
    1 * 25 g
       92,70 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 343,00 €
    343,00 €
    1 * 100 g
       343,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 710,00 €
    1 710,00 €
    1 * 500 g
       1 710,00 €

    Specifications:
    New Product
    New Product